Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ G60 Knuckle Coated High-Visibility Cut-Resistant Gloves
Click to view available options
Length (English):
8.4 in.
8.9 in.
9.3 in.
9.6 in.
10 in.
Length (Metric):
21.3 cm
22.6 cm
23.6 cm
24.3 cm
25.4 cm
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- EN 388 Level 3 / ANSI Level 2 Cut Resistance
- EN 388 Level 4 abrasion and Level 4 tear resistance
- Certified to meet the EN 420 standards
- Bright orange fiber aids in overall safety compliance and awareness of how close a hand is in proximity to potential dangers
- Made of ultra-high molecular weight polyethylene (UHMWPE) yarn with nylon and stretch fiber, which provides both cut resistance and a comfortable fit
- Polyurethane coating enhances dry and oil grip and hides dirt for longer wear
- Dark coating helps hide dirt for longer wear
- Each pair has a dedicated right and left glove for a snug fit
- Knuckle coating helps keep hands and gloves protected from liquids and oils to enforce compliance and improve glove longevity
Specifications
ANSI Abrasion Rating | ANSI 4 |
Certifications/Compliance (Europe) | EN 420 |
Color | Black, Orange |
Cut Resistance Rating | ANSI A3 |
Material | Dyneema™, Nylon, Lycra |
Product Type | Cut-resistant Gloves |
Size | Medium |
EN388 Standard | EN 388 |
For Use With (Application) | Metal fabrication, Glass handling, automotive assembly, stamping and assembly |
Glove Cuff Style | Knit |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction