Learn More
Kimberly-Clark Professional™ KleenGuard™ V40 Maxfire™ Safety Eyewear
Kimberly-Clark™ Professional KleenGuard™ V40 Maxfire™ Safety Eyewear feature 5-position click-to-fit temples for a personalized fit
$5.86 - $17.49
Specifications
Lens Type Anti-fog
Product Type Safety Eyewear
View More Specs
Products 3
Catalog Number Mfr. No. Frame Color Frame Type Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Frame Color Frame Type Lens Tint Price Quantity & Availability  
19-006-205
View Documents
Kimberly-Clark Professional™
38485
Brown Brown frame w/brown tips Smoke
Each for $17.49
Dozen of 12 EA for $210.00
Add to cart
19-006-207
View Documents
Kimberly-Clark Professional™
38494
Black Black frame w/red tips Amber
Each for $5.86
Dozen of 12 EA for $123.00
Add to cart
19-006-206
View Documents
Kimberly-Clark Professional™
38490
Brown Brown frame w/brown tips Indoor/Outdoor This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Ideal blend of comfort, style, flexibility and protection
  • Built-in comfort features including an adjustable nose pad and soft rubber inserts on the brow, temples and along the ear
  • Premium anti-scratch (EN 166 K Rating) and anti-fog (EN 166 N Rating) properties
  • Metal-free design (with screw-less hinges) helps minimize electrical transference risk
  • Meet ANSI Z87.1+ standards for high impact
  • Polycarbonate lenses provide 99.9% UVA / UVB / UVC protection
Specifications
Specifications
Anti-fog
Safety Eyewear