Learn More
Kimberly-Clark Professional™ KleenGuard™ V50 OTG™ Safety Glasses
Click to view available options
Lens Tint:
Clear
Indoor/Outdoor, Indoor/Outdoor Mirror
Smoke
Description
- Made with a nylon frame for strength, durability and comfort and fit over most prescription glasses
- See-through integrated side shields provide side protection without hindering peripheral vision
- Hard coated for scratch resistance and longer life
- Anti-fog coating for humid environment and temperature variances
- Meet ANSI Z87.1+ standards for high impact
- Polycarbonate lenses provide 99.9% UVA / UVB / UVC protection
Specifications
Certifications/Compliance | ANSI Z87.1 and CSA Z94.3 |
Lens Tint | Indoor/Outdoor, Indoor/Outdoor Mirror |
Product Type | OTG safety spectacles |
Frame Color | Black |
Material | PC |
Frame Material | PC |
UV Protection | 0.999 |