Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kimberly-Clark™ Professional W10 HSL 100 Passive Welding Helmet
Ideal for students, hobbyists and inspectors
Supplier: Kimberly-Clark™ Professional 14975
Description
Performance and value in an affordably priced helmet that meets safety requirements
- Extended front for throat protection from sparks, slag and other physical hazards
- Narrow shell design for work in tight space
- Equipped with a shade 10 polycarbonate filter
- Offers a clear, unobstructed view of the weld puddle
- Hard hat adaptable; ADF adaptable
- Size: 11.43 × 13.21cm (4.5 × 5.2 in.)
- Color: Black
Certifications
CE Certified
Compliance
- Meets ANSI Z87.1 – 2010 standards
- CSA compliant
Specifications
Black | |
HSL 100 passive welding helmet | |
4/Pk. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction