Learn More
Kimberly-Clark™ Professional KC200 Kimguard™ Sterilization Wraps Available on GSA/VA Contract for Federal Government customers only.
Soft, drapable wrap withstands tough handling
Manufacturer:  Kimberly-Clark™ Professional 68054
 View more versions of this product
Catalog No. 19-167-795
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Light Blue
54 x 54 in. (137.1 x 137.1cm)
54 in.
No
Yes
Steam and EtO
137.1cm
Low-lint
For linen towels, packs and gowns, light basin sets and single instruments and medical products
137.1cm
SMS Fabric
54 x 54 in. (137.1 x 137.1cm)
54 in.