Learn More
Kimberly-Clark™ Professional KC200 Kimguard™ Sterilization Wraps Available on GSA/VA Contract for Federal Government customers only.
Soft, drapable wrap withstands tough handling
Manufacturer:  Kimberly-Clark™ Professional 68040
 View more versions of this product
Catalog No. 19-065-704
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Light Blue
40 x 40 in. (101.6 x 101.6cm)
40 in.
No
Yes
40 in.
Low-lint
For linen towels, packs and gowns, light basin sets and single instruments and medical products
101.6cm
SMS Fabric, Polypropylene
Steam and EtO
101.6cm