Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimberly-Clark Professional™ KC400 Kimguard™ Sterilization Wraps

The fourth of six strength levels in the Kimguard line

Specifications

Color Medium Blue
Design Low-lint
For Use With (Application) Used for extra heavy instrument sets such as orthopedics and cardiovascular sets
Sterility Sterile
Latex Free Yes
View More Specs

Products 3
Catalog Number Mfr. No. Length (Metric) Width (Metric) Exterior Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Width (Metric) Exterior Price Quantity & Availability  
18-031-247
View Documents
Kimberly-Clark Professional™
68224
60.9 cm 60.9 cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
06-667-50
View Documents
Kimberly-Clark Professional™
68245
101.6 cm 101.6 cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-135-120
View Documents
Kimberly-Clark Professional™
68236
91.4 cm 91.4 cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Softer and stronger than ever
  • Delivers the barrier protection you expect from the leader in sterilization packaging
  • Used for extra heavy instrument sets such as orthopedics and cardiovascular sets
  • Color: Medium Blue

Order Info

Five additional strength levels (KC100, KC200, KC300, KC500 and KC600) available. Contact your Fisher Safety Customer Service Representative at 1-800-772-6733 for ordering information.

Specifications

Specifications

Medium Blue
Used for extra heavy instrument sets such as orthopedics and cardiovascular sets
Yes
Steam and EtO
Low-lint
Sterile
SMS Fabric
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit