Learn More
Kimberly-Clark™ Professional KC500 Kimguard™ Sterilization Wraps Available on GSA/VA Contract for Federal Government customers only.
Offers precisely right protection for specific needs
Manufacturer:  Kimberly-Clark™ Professional 68154
 View more versions of this product
Catalog No. 19-043-114
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Dark Blue
54 x 54 in. (137.1 x 137.1cm)
54 in.
No
Yes
54 in.
Low-lint
Used to package linen towels, packs, gowns, light basin sets and single instruments for sterilization
137.1cm
SMS Fabric
Steam and EO
137.1cm