Learn More
Kimberly-Clark™ Professional KC100 Kimguard One-Step™ Sterilization Wraps Available on GSA/VA Contract for Federal Government customers only.
Two sheets bonded together, making it possible to double wrap in a single step
Manufacturer:  Kimberly-Clark™ Professional 12745
 View more versions of this product
Catalog No. 19-041-985
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Light Blue
45 x 45 in. (114.3 x 114.3cm)
45 in.
No
Yes
45 in.
Low-lint
Light linen packs, linen gowns, towel packs, single basins and other lighter wrapping needs
114.3cm
SMS Fabric
Steam and EtO
114.3cm