Learn More
Kimberly-Clark™ Professional KC100 Kimguard One-Step™ Sterilization Wraps Available on GSA/VA Contract for Federal Government customers only.
Two sheets bonded together, making it possible to double wrap in a single step
Manufacturer:  Kimberly-Clark™ Professional 12748
 View more versions of this product
Catalog No. 19-041-986
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Light Blue
48 x 48 in. (121.92 x 121.92cm)
48 in.
No
Yes
48 in.
Low-lint
Light linen packs, linen gowns, towel packs, single basins and other lighter wrapping needs
121.92cm
SMS Fabric
Steam and EtO
121.92cm