Learn More
Kimberly-Clark™ Professional Kimguard One-Step™ KC200 Sterilization Wraps Available on GSA/VA Contract for Federal Government customers only.
Two sheets bonded together, making it possible to double wrap in single step
Manufacturer:  Kimberly-Clark™ Professional 62036
 View more versions of this product
Catalog No. 18-892-145
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Light Blue
Use to wrap parts, trays and kits prior to steam, gas or gamma irradiation
91.5cm
SMS Fabric
Steam and EtO
91.5cm
36 x 36 in. (91.5 x 91.5cm)
36 in.
Yes
Yes
36 in.