Learn More
Kimberly-Clark Professional™ Kimguard One-Step™ KC200 Sterilization Wraps
Two sheets bonded together, making it possible to double wrap in single step
Manufacturer:  Kimberly-Clark Professional™ 62048
 View more versions of this product
Catalog No. 19-065-708
Add to cart
Description
Description
KC200 is the second of six strength levels in the KIMGUARD ONE-STEP line. Use to wrap parts, trays and kits prior to steam, gas or gamma irradiation.
  • Patented design allows packages to be wrapped and opened in single step to save time
  • Imprinted with its name, size and lot code for convenience
  • Light blue SMS fabric
  • Non-sequential wrap
  • Latex free
  • Steam- and EtO-compatible
  • Recommended for lightweight packages
Specifications
Specifications
Safety
Light Blue
Use to wrap parts, trays and kits prior to steam, gas or gamma irradiation
121cm
Yes
48 in.
Sterile
48 x 48 in. (121 x 121cm)
48 in.
SMS Fabric
Steam and EtO
121cm