Kimberly-Clark™ Professional KC400 Kimguard One-Step™ Sterilization Wraps Available on GSA/VA Contract for Federal Government customers only.
Two sheets bonded together, making it possible to double wrap in single step
Manufacturer: Kimberly-Clark™ Professional 62236
 View more versions of this product
Catalog No. 19-043111
  Check Availability
Add to Cart
    Specifications
    Design Low-lint
    Dimensions (L x W) 36 x 36 in. (91.4 x 91.4cm)
    Sterile No
    Color Medium Blue
    Material SMS Fabric
    Latex Free Yes
    Sterilization Method Steam and EtO
  • Description & Specifications
Related Products
Product Content Feedback