Learn More
Kimberly-Clark™ Professional KC400 Kimguard One-Step™ Sterilization Wraps Available on GSA/VA Contract for Federal Government customers only.
Two sheets bonded together, making it possible to double wrap in single step
Manufacturer:  Kimberly-Clark™ Professional 62254
 View more versions of this product
Catalog No. 19-045375
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Medium Blue
54 x 54 in. (137.1 x 137.1cm)
SMS Fabric
Steam and EtO
Low-lint
No
Yes