Learn More
Kimberly-Clark™ Professional Kimtech Pure™ G3 Nonsterile Latex Ambidextrous Gloves
Flexibility minimizes fatigue
Manufacturer:  Kimberly-Clark™ Professional 56814
 View more versions of this product
Catalog No. 11-390-6B
  Check Availability
Add to cart
Alert:
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Specifications Description & Specifications
Specifications
Beaded/Rolled
Rubber
8.7mil
Natural
12 in.
Ambidextrous
Medium
Powder-free
Non-sterile
30.5cm