Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kimberly-Clark Professional™ Kimtech Science™ Kimwipes™ Delicate Task Wipers, 1-Ply
Shop All Kimberly-Clark Professional Products
Click to view available options
Dimensions (L x W):
11.2 x 12.3 in.
16.4 x 14.4 in.
8.2 x 4.4 in.
Packaging Quantity:
144 Wipes/Box, 15 Boxes/Cs.
198 Wipes/Box, 15 Boxes/Cs.
286 Wipes/Box, 30 Boxes/Cs.
286 Wipes/Box, 60 Boxes/Cs.
Description
Easily wipe up liquid and dust. Kimtech Science™ Kimwipes™ Delicate Task Wipers can handle a variety of delicate tasks. Their anti-static dispensing reduces lint and electrostatic discharge and controls usage.
- Single ply, light-duty wiper that can handle a variety of delicate tasks
- Extra low-lint wipers made from 100% virgin wood fibers
- Soft, nonabrasive cellulose-fiber wipers won't scratch most delicate surfaces
- Easily wipe up liquid and dust
- Absorb many times their own weight
- Handy one-at-a-time dispensing packs with special plastic guard to reduce airborne lint, keep contaminants out of box and reduce electrostatic discharge
Specifications
Specifications
| Product Type | Delicate Task Wipe |
| Length (English) | 8.2 in. |
| Length (Metric) | 20.83 cm |
| Width (English) | 4.4 in. |
| Width (Metric) | 11.17 cm |
| Material | Cellulose |
| Color | White |
| Disposable | Single-use |
| Dimensions (L x W) | 8.2 x 4.4 in. |
| For Use With (Application) | General-Purpose |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction