Kimberly-Clark™ Professional KleenGuard™ A10 Light Duty Shoe Covers

For a wide range of work situations, such as food processing, manufacturing facilities, hospitals, and construction sites

Manufacturer: Kimberly-Clark™ Professional 36811

 View more versions of this product

Catalog No. 19-057-428

  Check Availability
Add to Cart

    Specifications

    Size Universal
    Sole Type Low-skid
    Product Type Shoe Covers
    Material Spunbond Polypropylene
    For Use With (Application) Ideal to minimize the potential for tracking unwanted contaminants into sensitive areas and for cleanroom environments
    Closure Type Fully elastic
    Color Light Blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit