Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimberly-Clark Professional™ KleenGuard™ A20 Blue Coveralls

Catalog Number 190881931D
Click to view available options
Certifications/Compliance:
ANSI ISEA 101-1996
NFPA 99
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Designed for specific applications which feature ways of safeguarding and shielding itself and/or the user

  • Elastic Back, wrists and ankles
  • Attached hood
  • Disposable
  • Blue
  • Front closure: Zipper
  • Protective properties: Anti-static and particle protection

TRUSTED_SUSTAINABILITY
Size X-Large
Material SMS
Color Blue
Ankle Style Elastic
Cuff Style Elastic
Closure Type Front Zipper
Back Style Elastic
Certifications/Compliance NFPA 99
Description Microforce™ Barrier SMS Fabric
For Use With (Application) Laboratories, decontamination modesty garment, remediation, pesticides, heavy dust environments
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit