Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimberly-Clark Professional™ KleenGuard™ A20 Breathable Particle Protection Frocks
GREENER_CHOICE

Catalog Number 19169882
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4X-Large
Length (English):
40 in.
42 in.
44 in.
46 in.
Length (Metric):
102 cm
107 cm
112 cm
117cm
This item is not returnable. View return policy
This item is not returnable. View return policy

Protection against dry particulates and light liquid sprays

  • Snap front closure with snap at collar
  • Long length
  • Elastic wrists
  • No pockets
  • Passes NFPA 99 criteria for antistatic materials
  • Typical uses: Laboratories; Decontamination modesty garment; Remediation; Pesticides; Heavy dust environments
TRUSTED_SUSTAINABILITY
Size 4X-Large
Material SMS
Color White
Closure Type Snaps
For Use With (Application) Laboratories, decontamination modesty garment, remediation, pesticides, heavy dust environments
Certifications/Compliance NFPA 99
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit