Kimberly-Clark™ Professional KleenGuard™ A35 Coveralls
Manufacturer: Kimberly-Clark™ Professional 32602
 View more versions of this product
Catalog No. 19-457-124
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Certifications/Compliance ASTM F1670, ANSI ISEA 101-1996
    Material Fabric
    Includes Elastic wrist and ankle
    Size 4X-Large
    For Use With (Application) Liquid and particle protection
    Closure Type Zipper Front
  • Description & Specifications
Related Products
Product Content Feedback