Learn More
Kimberly-Clark™ Professional KleenGuard™ Eyewear: V40
Comfort, flexibility, and protection
Manufacturer:  Kimberly-Clark™ Professional 08155
 View more versions of this product
Catalog No. 19-167-777
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Impact-resistant lenses offer 99.9% UVA/UVB protection
  • Amber: For low-light environments where contrast is low; blocks out blue light to highlight and define shapes
  • Clear: For indoor applications; allows maximum visible light transmission
  • Smoke: For applications where eyestrain is a concern or bright, sunny days requiring maximum glare reduction
  • Silver Mirror: For outdoors where very bright sunlight and glare can cause eyestrain and fatigue
  • Indoor/Outdoor: For moving between indoor/shaded areas to direct sunlight and back
V40 Contour Model
  • Wraparound lens for better eye coverage
  • Soft, adjustable nose bridge
  • Comfortable, dual-molded rubber temples
  • Cushioned brow bar for additional comfort
Compliance
Meet ANSI™ Z87.1-2003 standards.
Specifications
Specifications
Uncoated
Dual-molded rubber
Silver Mirror
Impact-resistant