Learn More
Kimberly-Clark™ Professional KleenGuard™ Eyewear: V50
Deliversr comfort, flexibility, and protection
Manufacturer:  Kimberly-Clark™ Professional 0866100
 View more versions of this product
Catalog No. 19-168-320
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
With foam fog and particle shield. Anti-Fog. Features a proprietary, integrated fog and particle shield. Reduces fogging by 50 percent over traditional safety glasses.
  • Impact-resistant lenses offer 99.9% UVA/UVB protection
  • Amber: For low-light environments where contrast is low; blocks out blue light to highlight and define shapes
  • Clear: For indoor applications; allows maximum visible light transmission
  • Smoke: For applications where eyestrain is a concern or bright, sunny days requiring maximum glare reduction
  • Compliance
    Meet ANSI™ Z87.1-2003 standards.
Specifications
Specifications
Clear
Anti-fog