Kimberly-Clark™ Professional Nurses Surgical Masks Available on GSA/VA Contract for Federal Government customers only.
Three-layer construction protects nose and mouth with a comfortable fit
Manufacturer: Kimberly-Clark™ Professional 48210
 View more versions of this product
Catalog No. 19-166-994
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Filter Efficiency 97% (PFE), 96% (BFE)
    Color Blue
    Construction 3-ply construction
    Style Pleated
    Sterile No
    Pore Size 0.1 (PFE), 3 (BFE)μ
    Includes Tie closures
    Material Fabric
    Size Regular
  • Description & Specifications
Related Products
Product Content Feedback