Kimberly-Clark™ Professional PFE-Xtra™ Latex Exam Gloves
Ideal for procedures with risk of exposure to blood, body fluids, and chemicals
Manufacturer: Kimberly-Clark™ Professional 5050210
 View more versions of this product
Catalog No. 19-166-121
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
Alert
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
    Specifications
    Certifications AQL 1.5, FDA Class I Medical device, ASTM D 3578
    Glove Cuff Style Beaded
    Hand Ambidextrous
    Length (English) 12 in.
    Length (Metric) 30.5cm
    Material Natural Rubber
    Powdered No
    Size Medium
    Thickness 10.2mil
    Coating Material Textured
    Color Natural
  • Description & Specifications
Related Products
Product Content Feedback