Kimberly-Clark™ Professional PFR95™ N95 Particulate Filter Respirators Available on GSA/VA Contract for Federal Government customers only.

Pouch-style design provides large breathing area with limited facial contact

Manufacturer: Kimberly-Clark™ Professional 62126

 View more versions of this product

Catalog No. 18-999-4812

  Check Availability
Add to Cart

    Specifications

    Certifications NIOSH 42CFR84 N95
    Quantity 50 Pack
    Disposable Yes
    Filter Type N95 Filter
    For Use With (Application) Provides protection from blood borne pathogens and body fluids
    Size Regular
    Product Type Pouch
    Filter Efficiency 95% at 0.3μ
    Color White

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit