Kimberly-Clark™ Professional Procedure Gowns Get IT Item Available on GSA/VA Contract for Federal Government customers only.
Gowns are made with a protective fluid-resistant 3-layer SMS fabric and are recommended where there is the potential for low to moderate exposure to fluids or chemicals
Manufacturer: Kimberly-Clark™ Professional 69028
 View more versions of this product
Catalog No. 19-088-1698
  Check Availability
Add to Cart
    Specifications
    Closure Type Velcro™ neck
    Color Blue
    Size X-Large
    Cuff Style Knit
    Material SMS
  • Description & Specifications
Related Products
Product Content Feedback