Kimberly-Clark™ Professional Procedure Masks Available on GSA/VA Contract for Federal Government customers only.
22% lighter and cooler than average procedure mask
Manufacturer: Kimberly-Clark™ Professional 47090
 View more versions of this product
Catalog No. 19-166-980
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance CE
    Latex Free Yes
    Pore Size 0.1 (PFE), 3 (BFE)μ
    Width (Metric) 18cm
    Width (English) 7 in.
    Filter Efficiency ≥96% (BFE), ≥97% (PFE)
    Color White
    Construction 3-ply construction with white hydro-entangled Polyester binding, fully enclosed soft malleable Aluminum nose piece, Polypropylene, Cellulose, Polyurethane facing components
    Style Pleated
    Includes White tubular knitted Polyester, Polyurethane earloops
  • Description & Specifications
Related Products
Product Content Feedback