Kimberly-Clark™ Professional Procedure Masks
22% lighter and cooler than average procedure mask
Manufacturer: Kimberly-Clark™ Professional 47295
 View all options for this product
Catalog No. 19-166-991
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Color Pink and blue teddy bear print
    Construction 3-ply construction
    Includes Earloops
    Filter Efficiency ≥96% (BFE), ≥97% (PFE)
    Style Pleated
    Pore Size 0.1 (PFE), 3 (BFE)μ
    • Three-layer, pleat-style mask
    • Ear loops
    • Inner facing is light and breathable to reduce irritation
    • Outer facing resists fluid strike through
  • Description & Specifications
Related Products
Product Content Feedback