Learn More
Kimberly-Clark™ Professional Forest Green Nitrile Powder-Free Exam Gloves
Recyclable through RightCycle™ program
Manufacturer:  Kimberly-Clark™ Professional 43446
 View more versions of this product
Catalog No. 19-169-075
  Check Availability
Add to cart
Specifications
Description
  • Material: Nitrile
  • Thickness: 3.5mil
  • Ambidextrous
  • Textured fingertips
  • Beaded cuff, wet and dry grip
  • Free of natural rubber and latex-free
Description & Specifications
Specifications
Beaded/Rolled
Yes
Large
1.5 AQL exceeds the ASTM 2.5 standard for critical defects (pinholes)
Forest Green
Ambidextrous
Nitrile
2000/Cs.
Powder-free
Promotions
Promotions
  • Kimberly-Clark Corporation
    Buy one case of Kimberly-Clark™ exam gloves and receive two boxes at no additional cost. This is an NLSU exclusive offer. You must apply and qualify for the program to be eligible.
    12-31-2017