Learn More
Kimberly-Clark™ Professional Jackson Safety™ H10 Disposable Uncorded Earplugs Available on GSA/VA Contract for Federal Government customers only.
A unique combination of protection, comfort and performance from a high-quality hearing protection product
Manufacturer:  Kimberly-Clark™ Professional 67210
Catalog No. 19-168-895
Add to cart
Description
Description
  • Soft roll-down foam
  • Smooth bullet shaped for easy insertion
  • Ultra-smooth exterior for comfort
  • Individually packaged
  • Universal size
  • ANSI S3.19-1974
  • Color: Orange
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Universal
NRR 31; ANSI S3.19-1974
Orange
Soft roll-down foam