Learn More
Kimberly-Clark™ Professional Jackson Safety™ FlashTex™ 650 NPR Winter Liner
Ideal for very cold environments
Manufacturer:  Kimberly-Clark™ Professional 16766
Catalog No. 19-073-919
Add to cart
Description
Description
  • Cotton chinstrap Leather/Nomex™ tabs attach to hard hats
  • Longer nape for neck & lower forehead protection
  • Provides protection from electrical arc energy exposure
  • 100% Nomex™ IIIA outer shell maintains FR protection throughout life of product
  • 100% Proban™ cotton flame-resistant inner shell maintains flame resistance through 25 industrial washes or 50 domestic washes
Specifications
Specifications
Blue
Protection from electrical arc energy exposure, flame, very cold environments
Cotton/Nomex™
Hard hats