Kimberly-Clark™ Professional Jackson Safety™ G50 Mechanics Gloves with Palm and Finger Protection
Reinforced for work with rough materials and equipment
Manufacturer: Kimberly-Clark™ Professional 90269
 View more versions of this product
Catalog No. 19-812-854
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Size 10
    Offer extra protection and padding in the palm, fingertip and knuckle areas.
    • Breathable material between fingers for greater comfort
    • Padded palm provides excellent impact protection
    • Less bulky than leather
    Applications:
    Masonry, forging, stamping, injection molding.
  • Description & Specifications
Related Products
Product Content Feedback