Learn More
Kimberly-Clark™ Professional Jackson Safety™ G60 Cut-Resistant Gloves
For a higher level of hand protection in environments with a high risk for cutting injuries
Manufacturer:  Kimberly-Clark™ Professional 98248
 View more versions of this product
Catalog No. 19-168-886
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Ideal for use in paper making, glass cutting, metal stamping
  • Dyneema™ fabric construction
  • Level 5 cut resistance (EN388)
  • Smooth palm
  • Ambidextrous
  • Color: Purple
Description & Specifications
Specifications
Smooth
Ambidextrous
Dyneema™
Purple
Level 5
Glass/Metal handling, Paper making and automotive/aircraft assembly
10