Kimberly-Clark™ Professional Jackson Safety™ G60 Purple Nitrile™ Cut-Resistant Gloves

For industrial workers who need protection against hand cuts

Manufacturer: Kimberly-Clark™ Professional 97433

 View more versions of this product

Catalog No. 19-161-595

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Knit
    Hand Ambidextrous
    For Use With (Application) Glass/Metal handling, Paper making and automotive/aircraft assembly
    Material Dyneema™
    Size 10
    Coating Material Dotted textured
    Color Purple
    Cut Resistance Rating Level 3

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit