Kimberly-Clark™ Professional Jackson Safety™ Sentry III™ Cap
Low-riding nape strap prevents cap from slipping
Manufacturer: Kimberly-Clark™ Professional 14416
 View more versions of this product
Catalog No. 17-350-1C
  Check Availability
Add to Cart
    Specifications
    Construction Molded
    Material HDPE
    No. of Mounting Points Six
    Product Type Hard hat
    Certifications/Compliance Meets ANSI Z89.1 Class C, G, E
    Color Blue,Blue
    Suspension Type 6-Point Ratchet
  • Description & Specifications
Related Products
Product Content Feedback