Learn More
Kimberly-Clark Professional™ Jackson Safety™ V10 Expo™ Safety Eyewear Available on GSA/VA Contract for Federal Government customers only.
Polycarbonate lens with integral side protection and browguard
Manufacturer:  Kimberly-Clark Professional™ 22515
 View more versions of this product
Catalog No. 19-706-001
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Conventional, lightweight wraparound protection offers a wide field of vision
  • Four-base curve
  • Adjustable temple lengths for a custom fit to facial features
  • Durable, nylon frame
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Specifications
Smoke
Safety
Safety Glasses
Nylon
Safety glasses
Meets ANSI Z87.1-2010 standards.
Black
Polycarbonate