Kimberly-Clark™ Professional Kimcare™Antibacterial Skin Cleansers Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Kimberly-Clark™ Professional 92517

 View more versions of this product

Catalog No. 19-413-009

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Size (English) 0.13 gal.
    Size (Metric) 500mL
    Packaging Refill
    Quantity 18 Case
    Composition Triclosan 0.5%, Aloe and Provitamin B5
    Color Clear
    For Use With (Application) Protect against the spread of potentially harmful bacteria such as E. coli
    Fragrance Floral
    Product Type Antibacterial Skin Cleanser

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit