Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimberly-Clark Professional™ Kleenex™ General™ Gentle Skin Cleansers

Enriched with emolients to protect skin from dryness and irritation

Specifications

Boiling Point >100°C
Color Pink
For Use With (Application) Used to cleanse or remove light soil on Gentle Skin
Fragrance Floral
View More Specs

Products 1
Catalog Number Mfr. No. Capacity Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Price Quantity & Availability  
19-413-005 View Documents Kimberly-Clark Professional™
91552
1000mL Case of 6 for $109.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • Mild, all purpose moisturising skin cleanser
  • Ideal for public facilities including hospitality, office buildings, hospitals, restaurants, etc.
  • pH balanced resulting in healthy hands
  • Pleasant floral fragrance

Specifications

Specifications

>100°C
Pink
Used to cleanse or remove light soil on Gentle Skin
Floral
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit