Kimberly-Clark™ Professional Kimtech Pure™ A8 Shoe Covers Available on GSA/VA Contract for Federal Government customers only.

For clean manufacturing environments

Manufacturer: Kimberly-Clark™ Professional 39370

 View more versions of this product

Catalog No. 19-169-650

  Check Availability
Add to Cart

    Specifications

    Size Small, Medium
    Sole Type Extra grip, seamless
    Product Type Shoe Covers
    Material Spunbond, Polyethylene, Polyester
    For Use With (Application) Ideal for clean manufacturing environment
    Closure Type Elastic toe and heel
    Latex Free Yes
    Thread Color White
    Color Blue
    Construction Spunbond, Polyethylene Film Laminate, Polyester Thread

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit