Learn More
Kimberly-Clark Professional™ Kimtech Pure™ G3 EvT Nitrile Gloves
For ISO Class 3 and higher environments
Manufacturer:  Kimberly-Clark Professional™ 38701
 View more versions of this product
Catalog No. 19-169-451
Add to cart
Description
Description
  • A natural rubber latex-free glove that reduces the risk of Type I allergic reactions
  • Low particles and extractables—designed specifically for use in cleanroom manufacturing environments
  • Cleaned to meet the needs of the critical cleanroom environment
  • Textured and tacky surface provides good grip
  • AQL 1.5 for pinholes
  • Ambidextrous
  • 12 in. length
  • 5mil thickness
  • Can be recycled in the RightCycle™ by Kimberly-Clark Professional program
Specifications
Specifications
ISO Class 3
Ambidextrous
Small
White
30.48cm
Textured, tacky
Nitrile
5 mil
12 in.