Kimberly-Clark™ Professional Kimtech Pure™ G3 and G5 STERLING™ Nitrile Gloves

Combine the protection of nitrile with the sensitivity of latex

Manufacturer: Kimberly-Clark™ Professional 98187

 View more versions of this product

Catalog No. 17-986-010

  Check Availability
Add to Cart

    Specifications

    Exterior Finish Textured
    Glove Cuff Style Beaded
    Hand Ambidextrous
    Length (English) 12 in.
    Length (Metric) 30.48cm
    Powdered No
    Sterile No
    Thickness 4.0mil
    Classification ISO Class 5
    Material Nitrile
    Size Large
    Color Gray

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit