Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimberly-Clark™ Professional Kimtech Pure™ G3 Nitrile Gloves

Manufacturer-suggested ISO Class 3 or higher

Manufacturer:  Kimberly-Clark™ Professional 56877

 View more versions of this product

Catalog No. 19-048-591C



  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

ISO Class 3
Beaded/Rolled
Nitrile
Medium
EN374-3:2003, IEST-RP-CC005 EN374-1:2003, AQL 1.5 / EN374-2, ISO16604, CE directive 89/686/EEC
Blue
30.48cm
Textured
Ambidextrous
Virus
5.1mil
Nitrile Gloves
12 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit