Kimberly-Clark™ Professional Kimtech Pure™ G3 NXT™ Nitrile Gloves

Contains no natural rubber latex proteins. Kimtech Pure™ G3 NXT™ Nitrile Gloves are suggested for ISO Class 3 or higher cleanroom environments.

Manufacturer: Kimberly-Clark™ Professional 62994

 View more versions of this product

Catalog No. 19-059-326E

  Check Availability
Add to Cart

    Specifications

    Exterior Finish Textured
    Glove Cuff Style Beaded
    Length (English) 12 in.
    Length (Metric) 30.48cm
    Material Nitrile
    Powdered No
    Thickness 5.1mil
    Classification Class 100
    Hand Ambidextrous
    Size X-Large
    Color White

    • Textured fingertips
    • Beaded cuff for easy donning
    • Contain no natural rubber latex
    • Ambidextrous
    • Thickness: 6.2mil at fingertips
    • Length: 12 in. (30.5cm)
    • White

     

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit