Learn More
Kimberly-Clark Professional™ Kimtech™ Pure G5 Latex 12 in. Cleanroom Gloves
G5 non-sterile 12 in. latex gloves provide enhanced tactile sensitivity and performance. Recommended for Class 100/ISO 5 or higher critical environments.
$41.64
Specifications
Certifications/Compliance ASTM D3578
Cleanroom Class Class 100/ISO 5
AQL 1.5
Thickness (English) 8 mil
Thickness (Metric) 0.20 mm
View More Specs
Alert:
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
11-390-5A
View Documents Promotion Details
Kimberly-Clark Professional™
56809
Small
Pack of 100 for $41.64
Case of 10 PK for $416.39
Add to cart
11-390-5B
View Documents Promotion Details
Kimberly-Clark Professional™
56810
Medium
Pack of 100 for $41.64
Case of 10 PK for $416.39
Add to cart
11-390-5C
View Documents Promotion Details
Kimberly-Clark Professional™
56811
Large
Pack of 100 for $41.64
Case of 10 PK for $416.39
Add to cart
11-390-5D
View Documents Promotion Details
Kimberly-Clark Professional™
56812
X-Large
Pack of 100 for $41.64
Case of 10 PK for $416.39
Add to cart
Description
Description
  • Low in natural rubber proteins
  • Textured palm and fingers
  • Beaded cuff adds strength and makes donning easier
  • Ambidextrous
  • Thickness: 8.3 mil
  • Length: 12 in. (30 cm)
Specifications
Specifications
ASTM D3578
1.5
0.20 mm
30.5 cm
Class 100/ISO 5
8 mil
12 in.
No Texture