Kimberly-Clark™ Professional Kimtech Pure™ G5 Nitrile Gloves

Manufacturer-suggested ISO Class 5

Manufacturer: Kimberly-Clark™ Professional 56861

 View more versions of this product

Catalog No. 11-390-13C

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Beaded
    Exterior Finish Bisque
    Hand Ambidextrous
    Length (English) 10 in.
    Length (Metric) 25.4cm
    Material Synthetic Nitrile Polymer
    Powdered No
    Size Large
    Thickness 5.1mil
    Color Blue
    Classification ISO Class 5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit