Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ A10 Light-Duty Coveralls
The right combination of price and protection for nonhazardous environments
Manufacturer: Kimberly-Clark Professional™ 10608
This item is not returnable.
View return policy
Description
- Lightweight, breathable fabric keeps workers cool and comfortable
- Zipper front
- Elastic wrists
- Also available with attached hood and boots
- White
Compliance
Meets ANSI™/ISEA™ 101-1996 standards.
Specifications
Front zipper | |
Polypropylene | |
ANSI/ISEA 101-1996 | |
Elastic | |
Food processing, general maintenance, manufacturing | |
Serged |
Light Duty Coverall | |
Attached boots | |
White | |
With attached hood and boots; Breathable SMS Polypropylene | |
Hooded | |
X-Large |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title