Kimberly-Clark™ Professional KleenGuard™ A10 Light-Duty Coveralls Available on GSA/VA Contract for Federal Government customers only.

The right combination of price and protection for nonhazardous environments

Manufacturer: Kimberly-Clark™ Professional 10606

 View more versions of this product

Catalog No. 19-812-880

  Check Availability
Add to Cart

    Specifications

    Product Type Light Duty Coverall
    Certifications/Compliance ANSI ISEA 101-1996
    Material Polypropylene
    Seam Type Serged
    Color White
    Size Large
    For Use With (Application) Food processing, general maintenance, manufacturing
    Closure Type Zipper Front
    Construction Breathable SMS Polypropylene
    Includes With attached hood and boots
    Cuff Style Elastic

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit