Kimberly-Clark™ Professional KleenGuard™ A10 Light-Duty Lab Coat

Lightweight, breathable fabric keep workers cool and comfortable

Manufacturer: Kimberly-Clark™ Professional 40106

 View more versions of this product

Catalog No. 19-057-416D

  Check Availability
Add to Cart

    Specifications

    Size 3X-Large
    Disposable Yes
    Closure Type Snap Front
    Cuff Style Elastic
    Material Fabric, Polypropylene
    For Use With (Application) Suited for food processing, general maintenance, manufacturing
    Certifications/Compliance ANSI ISEA 101-1996
    Color White

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit