Learn More
Kimberly-Clark Professional™ KleenGuard™ A20 Breathable Particle Protection Coveralls: Elastic Back, Wrist and Ankles with Attached Hood and Boots
GREENER_CHOICE
Patented Reflex™ design is 7.5 times less likely to rip out than ANSI™ minimums — provides 12% more chest room and 6% longer body length
$400.71 - $458.09
Specifications
Ankle Style Elastic, Attached boots
Back Style Elastic
Certifications/Compliance NFPA 99
Closure Type Front zipper
Color White
View More Specs
Alert:
Not for use near heat, flames or flammable environments
Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-981-48D
GREENER_CHOICE
View Documents
Kimberly-Clark Professional™
49126
3X-Large
Case of 20 for $400.71
Add to cart
17-981-48E
GREENER_CHOICE
View Documents
Kimberly-Clark Professional™
49122
Medium
Case of 24 for $458.09
Add to cart
17-981-48C
GREENER_CHOICE
View Documents
Kimberly-Clark Professional™
49125
2X-Large
Case of 24 for $457.18
Add to cart
17-981-48A
GREENER_CHOICE
View Documents
Kimberly-Clark Professional™
49123
Large
Case of 24 for $458.09
Add to cart
17-981-48B
GREENER_CHOICE
View Documents
Kimberly-Clark Professional™
49124
X-Large
Case of 24 for $458.09
Add to cart
19-169-900
GREENER_CHOICE
View Documents
Kimberly-Clark Professional™
49127
4X-Large
Case of 20 for $372.81
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
For use in laboratories, remediation, decontamination modesty garment, pesticides, and heavy dust environments. Elastic Back, Wrist and Ankles with Attached Hood and Boots.
  • Provide general protection against many particles
  • Breathable patented Microforce™ barrier SMS fabric passes NFPA 99 for antistatic materials
  • Offers high abrasion resistance and is low linting
  • Withstand multiple wearings
  • Air and moisture vapor can pass through to aid wearer comfort
  • Triple-stitched serged seams
  • White
Specifications
Specifications
Elastic, Attached boots
NFPA 99
White
SMS
Elastic
Front zipper
Elastic
Laboratories, decontamination modesty garment, remediation, pesticides, heavy dust environments