Kimberly-Clark™ Professional KleenGuard™ A20 Breathable Particle Protection Coveralls: Elastic Back, Wrists and Ankles with Attached Hood

Patented Reflex™ design is 7.5 times less likely to rip out than ANSI™ minimums — provides 12% more chest room and 6% longer body length

Manufacturer: Kimberly-Clark™ Professional 49114

 View more versions of this product

Catalog No. 17-981-47B

  Check Availability
Add to Cart

Alert

Not for use near heat, flames or flammable environments


    Specifications

    Certifications/Compliance NFPA 99
    Material SMS Fabric
    Color White
    For Use With (Application) Laboratories, decontamination modesty garment, remediation, pesticides, heavy dust environments
    Construction Microforce™ Barrier SMS Fabric
    Includes Attached hood
    Size X-Large
    Closure Type Zipper Front
    Back Style Elastic
    Ankle Style Elastic
    Cuff Style Elastic

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit